Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRFN1
Entrez Gene ID
102139971
Gene Name
leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases
-
Chromosome Location
Chr.19: 39803051 - 39817174 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589171.2 XP_005589228.1 3 - 4 3627 87

View Full Product Details