Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMX2
Entrez Gene ID
102137050
Gene Name
H6 family homeobox 2
Gene Aliases
-
Chromosome Location
Chr.9: 122422167 - 122430441 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566669.2 XP_005566726.1 2 - 3 347 100
XM_005566670.2 XP_005566727.1 2 - 3 329 100

View Full Product Details