Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C2H3orf38
Entrez Gene ID
102146261
Gene Name
chromosome 2 open reading frame, human C3orf38
Gene Aliases
-
Chromosome Location
Chr.2: 180012350 - 180022293 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005548395.2 XP_005548452.1 2 - 3 760 93

View Full Product Details