Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD9A
Entrez Gene ID
101926088
Gene Name
RAD9 checkpoint clamp component A
Gene Aliases
-
Chromosome Location
Chr.14: 7055383 - 7142517 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284976.1 NP_001271905.1 10 - 11 1065 95
XM_015434157.1 XP_015289643.1 5 - 6 954 95
XM_015434158.1 XP_015289644.1 10 - 11 2292 95
XM_015434159.1 XP_015289645.1 5 - 6 912 95
XM_015434160.1 XP_015289646.1 10 - 11 2254 95
XM_015434162.1 XP_015289648.1 3 - 4 744 95
XM_015434163.1 XP_015289649.1 3 - 4 702 95
GenBank mRNA AB169413.1 - 10 - 11 1065 95

View Full Product Details