Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEURL1B
Entrez Gene ID
102115928
Gene Name
neuralized E3 ubiquitin protein ligase 1B
Gene Aliases
-
Chromosome Location
Chr.6: 172270593 - 172320773 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558531.2 XP_005558588.1 2 - 3 747 70

View Full Product Details