Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN2
Entrez Gene ID
102145924
Gene Name
claudin 2
Gene Aliases
SP82
Chromosome Location
Chr.X: 104043559 - 104121296 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005594292.2 XP_005594349.1 2 - 3 1051 89

View Full Product Details