Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSBP1L1
Entrez Gene ID
102127974
Gene Name
heat shock factor binding protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.18: 324618 - 342269 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005586359.2 XP_005586416.1 2 - 3 265 67
XM_015439588.1 XP_015295074.1 2 - 3 272 67

View Full Product Details