Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102141134
Entrez Gene ID
102141134
Gene Name
signal-regulatory protein beta-1
Gene Aliases
SIRPB1
Chromosome Location
Chr.10: 60996000 - 61019936 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005568541.2 XP_005568598.1 4 - 5 1155 60

View Full Product Details