Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FJX1
Entrez Gene ID
102132930
Gene Name
four-jointed box kinase 1
Gene Aliases
-
Chromosome Location
Chr.14: 31807142 - 31809437 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005596035.2 XP_005596092.2 1 - 1 421 92

View Full Product Details