Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR2J
Entrez Gene ID
102125950
Gene Name
RNA polymerase II subunit J
Gene Aliases
-
Chromosome Location
Chr.3: 46812940 - 46818752 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005549315.2 XP_005549372.1 2 - 3 311 108
XM_005549316.2 XP_005549373.1 2 - 3 288 108

View Full Product Details