Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CABP7
Entrez Gene ID
102137830
Gene Name
calcium binding protein 7
Gene Aliases
-
Chromosome Location
Chr.10: 21745016 - 21756875 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005567649.2 XP_005567706.1 3 - 4 448 59

View Full Product Details