Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddr1
Entrez Gene ID
12305
Gene Name
discoidin domain receptor family, member 1
Gene Aliases
6030432F18, AI323681, CD167a, Cak, Nep, PTK3A
Chromosome Location
Chr.17: 35681567 - 35704629 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198831.1 NP_001185760.1 3 - 4 278 68
NM_001198833.1 NP_001185762.1 3 - 4 397 68
NM_007584.2 NP_031610.1 3 - 4 453 68
NM_172962.1 NP_766550.1 3 - 4 453 68
GenBank mRNA AK031442.1 - 3 - 4 356 68
AK143088.1 - 3 - 4 346 68
BC006836.1 - 3 - 4 359 68
BC037108.1 - 4 - 5 532 68
BC065998.1 - 3 - 4 289 68
CK790776.1 - 3 - 4 278 68
L57509.1 - 3 - 4 301 68

View Full Product Details