The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007870.3 | NP_031896.3 | 6 - 7 | 913 | 78 |
GenBank mRNA | AF047355.1 | - | 6 - 7 | 888 | 78 |
AK156109.1 | - | - | 746 | 78 | |
AK171843.1 | - | 6 - 7 | 746 | 78 | |
AK172254.1 | - | 6 - 7 | 1306 | 78 | |
BC012671.1 | - | 6 - 7 | 866 | 78 | |
U76110.1 | - | 6 - 7 | 722 | 78 |