The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267710.1 | NP_001254639.1 | 8 - 9 | 1158 | 81 |
NM_001267711.1 | NP_001254640.1 | 8 - 9 | 1153 | 81 | |
NM_007908.4 | NP_031934.1 | 8 - 9 | 1242 | 81 | |
GenBank mRNA | AK028524.1 | - | 8 - 9 | 1219 | 81 |
AK080915.1 | - | 8 - 9 | 1159 | 81 | |
AK135891.1 | - | 6 - 7 | 584 | 81 | |
AK136015.1 | - | 8 - 9 | 1154 | 81 | |
BC003433.1 | - | 8 - 9 | 1190 | 81 | |
BC055361.1 | - | 8 - 9 | 1153 | 81 | |
U93848.1 | - | 7 - 8 | 896 | 81 |