The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008465.5 | NP_032491.2 | 10 - 11 | 1148 | 96 |
GenBank mRNA | AK028259.1 | - | 10 - 11 | 1149 | 96 |
AK028307.1 | - | 10 - 11 | 1167 | 96 | |
AK154800.1 | - | 10 - 11 | 1166 | 96 | |
AK169311.1 | - | 10 - 11 | 1171 | 96 | |
AK169348.1 | - | 10 - 11 | 1171 | 96 | |
BC006771.1 | - | 10 - 11 | 1133 | 96 | |
U20619.1 | - | 10 - 11 | 1153 | 96 | |
U34228.1 | - | 9 - 10 | 997 | 96 |