The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029837.2 | NP_001025008.2 | 5 - 6 | 878 | 65 |
NM_001164049.1 | NP_001157521.1 | 5 - 6 | 905 | 65 | |
NM_001164050.1 | NP_001157522.1 | 5 - 6 | 905 | 65 | |
NM_001164051.1 | NP_001157523.1 | 5 - 6 | 929 | 65 | |
NM_001164052.1 | NP_001157524.1 | 3 - 4 | 632 | 65 | |
NM_008840.3 | NP_032866.2 | 6 - 7 | 1080 | 65 | |
GenBank mRNA | AK040867.1 | - | 5 - 6 | 840 | 65 |
AK149935.1 | - | 6 - 7 | 979 | 65 | |
AK150649.1 | - | 5 - 6 | 806 | 65 | |
AK151482.1 | - | 5 - 6 | 806 | 65 | |
AK152661.1 | - | 5 - 6 | 806 | 65 | |
AK153304.1 | - | 5 - 6 | 840 | 65 | |
AK155835.1 | - | 5 - 6 | 791 | 65 | |
AK171025.1 | - | 5 - 6 | 888 | 65 | |
AK172222.1 | - | 5 - 6 | 782 | 65 | |
BC035203.1 | - | 5 - 6 | 895 | 65 | |
CT010349.1 | - | 3 - 4 | 600 | 65 | |
U86587.1 | - | 3 - 4 | 600 | 65 |