Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sema3b
Entrez Gene ID
20347
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B
Gene Aliases
SemA, Semaa, sema5, semaV
Chromosome Location
Chr.9: 107597674 - 107609303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042779.2 NP_001036244.1 17 - 18 2050 68
NM_001291537.1 NP_001278466.1 17 - 18 2314 68
NM_001291538.1 NP_001278467.1 17 - 18 2308 68
NM_001291539.1 NP_001278468.1 16 - 17 1991 68
NM_009153.3 NP_033179.2 15 - 16 1860 68
NR_111986.1 15 - 16 1803 68
GenBank mRNA AK014333.1 - 15 - 16 1739 68
AK033597.1 - 16 - 17 2001 68
AK037034.1 - 17 - 18 2039 68
AK043634.1 - 16 - 17 1944 68
AK048364.1 - 16 - 17 2272 68
AK148232.1 - 17 - 18 2011 68
AK205569.1 - 1 - 2 91 68
BB116052.2 - 4 - 5 315 68
BC090669.1 - 16 - 17 1971 68
BC150758.1 - 14 - 15 1706 68
X85990.1 - 15 - 16 1857 68

View Full Product Details