Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpini1
Entrez Gene ID
20713
Gene Name
serine (or cysteine) peptidase inhibitor, clade I, member 1
Gene Aliases
AI837402, Ns, PI-12, PI12, Spi17
Chromosome Location
Chr.3: 75557503 - 75642523 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009250.2 NP_033276.1 7 - 8 1188 85
GenBank mRNA AJ001700.1 - 7 - 8 1157 85
AK032152.1 - 7 - 8 1177 85
AK133770.1 - 7 - 8 86
BC006776.1 - 7 - 8 1153 85

View Full Product Details