The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009850.2 | NP_033980.1 | 1 - 2 | 166 | 79 |
GenBank mRNA | AK134049.1 | - | 1 - 2 | 99 | 79 |
AK153980.1 | - | 1 - 2 | 98 | 79 | |
AK153987.1 | - | 1 - 2 | 85 | 79 | |
BC027528.1 | - | 1 - 2 | 97 | 79 | |
EF474453.1 | - | 1 - 2 | 55 | 79 | |
GU456630.1 | - | 1 - 2 | 55 | 79 | |
Y00635.1 | - | 1 - 2 | 194 | 79 |