Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpd2
Entrez Gene ID
14571
Gene Name
glycerol phosphate dehydrogenase 2, mitochondrial
Gene Aliases
AA408484, AI448216, AU021455, AW494132, GPDH, Gdm1, Gpd-m, Gpdh-m, TISP38
Chromosome Location
Chr.2: 57237678 - 57370719 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145820.1 NP_001139292.1 10 - 11 1622 60
NM_010274.3 NP_034404.3 10 - 11 1497 60
GenBank mRNA AK034353.1 - 10 - 11 1406 60
AK144716.1 - 10 - 11 1623 60
AK151189.1 - 10 - 11 1410 60
AK167152.1 - 10 - 11 1435 60
BC021359.1 - 10 - 11 1591 60
D50430.1 - 10 - 11 1399 60
U60987.1 - 10 - 11 1433 60

View Full Product Details