The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010892.3 | NP_035022.2 | 7 - 8 | 1289 | 89 |
GenBank mRNA | AF007247.1 | - | 7 - 8 | 1203 | 89 |
AF013166.1 | - | 7 - 8 | 1173 | 89 | |
AK147072.1 | - | 7 - 8 | 1265 | 89 | |
AK164467.1 | - | 7 - 8 | 1232 | 89 | |
AK169330.1 | - | 8 - 9 | 1180 | 89 | |
AK170179.1 | - | 7 - 8 | 1209 | 89 | |
AK171277.1 | - | 7 - 8 | 1209 | 89 | |
AK171387.1 | - | 7 - 8 | 1209 | 89 | |
BC010302.1 | - | 7 - 8 | 1211 | 89 | |
BC011316.1 | - | 7 - 8 | 1209 | 89 | |
BC057576.1 | - | 7 - 8 | 1229 | 89 | |
U95610.1 | - | 7 - 8 | 1200 | 89 |