The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011076.2 | NP_035206.2 | 24 - 25 | 3207 | 80 |
GenBank mRNA | AK144907.1 | - | 6 - 7 | 936 | 80 |
AY864315.1 | - | 23 - 24 | 3070 | 80 | |
M24417.1 | - | 18 - 19 | 2556 | 80 | |
M33581.1 | - | 24 - 25 | 3206 | 80 |