The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083935.1 | NP_001077404.1 | 8 - 9 | 1114 | 110 |
NM_001083936.1 | NP_001077405.1 | 7 - 8 | 1147 | 110 | |
NM_011199.2 | NP_035329.2 | 9 - 10 | 1116 | 110 | |
GenBank mRNA | BC013446.1 | - | 9 - 10 | 1093 | 110 |
BC051981.1 | - | 7 - 8 | 1018 | 110 | |
X78936.1 | - | 7 - 8 | 893 | 110 |