Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc4a1
- Entrez Gene ID
-
20533
- Gene Name
- solute carrier family 4 (anion exchanger), member 1
- Gene Aliases
-
Ae1, CD233, Empb3, l11Jus51
- Chromosome Location
-
Chr.11: 102348820 - 102365281 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK171662.1 |
- |
- |
3144 |
65 |
AK171845.1 |
- |
- |
3144 |
65 |
- Gene Symbol
- Syk
- Entrez Gene ID
-
20963
- Gene Name
- spleen tyrosine kinase
- Gene Aliases
-
Sykb
- Chromosome Location
-
Chr.13: 52582329 - 52648792 on Build GRCm38
View Full Product Details