The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011596.5 | NP_035726.2 | - | 1161 | 141 |
GenBank mRNA | AB022323.1 | - | - | 1127 | 141 |
AF218252.1 | - | - | 1146 | 141 | |
AK155055.1 | - | - | 1138 | 141 | |
AK170122.1 | - | - | 1415 | 141 | |
BC018369.1 | - | - | 1114 | 141 | |
BC108991.1 | - | - | 1058 | 141 | |
BC108992.1 | - | - | 1058 | 141 | |
BC112905.1 | - | - | 1121 | 141 | |
M31226.1 | - | - | 1057 | 141 | |
X55184.1 | - | - | 1058 | 141 |