The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011659.2 | NP_035789.1 | 1 - 2 | 309 | 64 |
GenBank mRNA | BC065782.1 | - | 1 - 2 | 148 | 64 |
BC139239.1 | - | 1 - 2 | 179 | 64 | |
BC139266.1 | - | 1 - 2 | 179 | 64 | |
Z21674.1 | - | 1 - 2 | 130 | 64 |