The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161824.1 | NP_001155296.1 | 4 - 5 | 763 | 94 |
NM_013604.2 | NP_038632.2 | 4 - 5 | 766 | 94 | |
GenBank mRNA | AK086135.1 | - | 4 - 5 | 359 | 94 |
AK159316.1 | - | 3 - 4 | 453 | 94 | |
AK163982.1 | - | 4 - 5 | 334 | 94 | |
AK165547.1 | - | 4 - 5 | 341 | 94 | |
AK165923.1 | - | 4 - 5 | 339 | 94 | |
BB662550.1 | - | 4 - 5 | 358 | 94 | |
BC023071.1 | - | 4 - 5 | 337 | 94 | |
L36962.1 | - | 4 - 5 | 336 | 94 |