The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013657.5 | NP_038685.3 | 7 - 8 | 841 | 119 |
GenBank mRNA | AK140348.1 | - | - | 1377 | 119 |
BC066852.1 | - | 7 - 8 | 841 | 119 | |
X85994.1 | - | 7 - 8 | 793 | 119 |