Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sema3c
Entrez Gene ID
20348
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C
Gene Aliases
1110036B02Rik, SemE, Semae
Chromosome Location
Chr.5: 17574816 - 17730268 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013657.5 NP_038685.3 15 - 16 1836 77
GenBank mRNA AK034239.1 - 15 - 16 1895 77
AK077231.1 - 4 - 5 959 77
AK140348.1 - - 2372 77
BC066852.1 - 15 - 16 1836 77
X85994.1 - 15 - 16 1788 77

View Full Product Details