The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015826.5 | NP_056641.2 | 4 - 5 | 1111 | 74 |
GenBank mRNA | AB221653.1 | - | 4 - 5 | 963 | 74 |
AF202778.1 | - | 4 - 5 | 1098 | 74 | |
AL133300.1 | - | 4 - 5 | 1114 | 74 | |
AY169785.1 | - | 3 - 4 | 830 | 74 | |
AY169786.1 | - | 4 - 5 | 1114 | 74 | |
AY169787.1 | - | 4 - 5 | 1114 | 74 | |
AY943925.1 | - | 4 - 5 | 653 | 74 | |
BC125474.1 | - | 4 - 5 | 1074 | 74 |