The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163336.1 | NP_001156808.1 | 7 - 8 | 813 | 62 |
NM_001163337.1 | NP_001156809.1 | 7 - 8 | 813 | 62 | |
NM_016745.3 | NP_058025.2 | 7 - 8 | 813 | 62 | |
GenBank mRNA | AK089487.1 | - | 7 - 8 | 813 | 62 |
AK155904.1 | - | 7 - 8 | 814 | 62 | |
BC017639.1 | - | 7 - 8 | 727 | 62 | |
BC026147.1 | - | 7 - 8 | 759 | 62 | |
U49393.1 | - | 7 - 8 | 674 | 62 | |
U49394.1 | - | 7 - 8 | 674 | 62 |