The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276704.1 | NP_001263633.1 | 1 - 2 | 147 | 72 |
NM_016813.2 | NP_058093.2 | 1 - 2 | 161 | 72 | |
GenBank mRNA | AF093140.1 | - | 1 - 2 | 137 | 72 |
AK077644.1 | - | 1 - 2 | 118 | 72 | |
AK079583.1 | - | 1 - 2 | 148 | 72 | |
AK086527.1 | - | 1 - 2 | 158 | 72 | |
AK088634.1 | - | 1 - 2 | 118 | 72 | |
AK133883.1 | - | 1 - 2 | 150 | 72 | |
AK143640.1 | - | - | 148 | 72 | |
AK167518.1 | - | 1 - 2 | 72 | ||
BC005594.1 | - | 1 - 2 | 161 | 72 |