Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nxf1
Entrez Gene ID
53319
Gene Name
nuclear RNA export factor 1
Gene Aliases
Mex67, Mvb1, Tap
Chromosome Location
Chr.19: 8757103 - 8770910 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276704.1 NP_001263633.1 17 - 18 3450 82
NM_016813.2 NP_058093.2 18 - 19 1699 82
GenBank mRNA AF093140.1 - 18 - 19 1675 82
AK052937.1 - 8 - 9 866 82
AK077644.1 - 18 - 19 1656 82
AK079583.1 - 18 - 19 1688 82
AK086527.1 - 17 - 18 3461 82
AK088634.1 - 17 - 18 3421 82
AK133883.1 - 18 - 19 1688 82
AK143640.1 - - 3451 82
AK167518.1 - 18 - 19 1687 82
BC005594.1 - 18 - 19 1699 82

View Full Product Details