Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cftr
Entrez Gene ID
12638
Gene Name
cystic fibrosis transmembrane conductance regulator
Gene Aliases
AW495489, Abcc7
Chromosome Location
Chr.6: 18170262 - 18322774 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021050.2 NP_066388.1 25 - 26 4257 97
GenBank mRNA M60493.1 - 25 - 26 4258 97
M69298.1 - 25 - 26 4256 97

View Full Product Details