Product Details
- Assay Reference Genome
Location
Chr.16:14447646 on build GRCm38
- Cytoband
- 16A1
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 17250
- Gene Symbol
- Abcc1
- Gene Name
- ATP-binding cassette, sub-family C (CFTR/MRP), member 1
- Gene Aliases
- Abcc1a, Abcc1b, MRP, Mdrap, Mrp1
- Location
- Chr.16:14361521-14474879 on build GRCm38
- Assay Gene Location
- Overlaps Exon 18 - Intron 18
View Full Product Details