The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008909.2 | NP_032935.2 | 2 - 3 | 302 | 55 |
GenBank mRNA | AF126834.1 | - | 2 - 3 | 242 | 55 |
AK142562.1 | - | 2 - 3 | 297 | 55 | |
BC057152.1 | - | 2 - 3 | 231 | 55 | |
CJ056283.1 | - | 2 - 3 | 302 | 55 |