The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159401.1 | NP_001152873.1 | 5 - 6 | 577 | 54 |
NM_001159402.1 | NP_001152874.1 | 5 - 6 | 576 | 54 | |
NM_009477.3 | NP_033503.2 | 5 - 6 | 604 | 54 | |
GenBank mRNA | AK136474.1 | - | 5 - 6 | 457 | 54 |
BC036559.1 | - | 5 - 6 | 478 | 54 | |
CA493594.1 | - | - | 614 | 54 | |
D44464.1 | - | 5 - 6 | 605 | 54 |