The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009510.2 | NP_033536.2 | 11 - 12 | 1481 | 80 |
GenBank mRNA | AK002766.1 | - | 11 - 12 | 1455 | 80 |
AK035271.1 | - | 7 - 8 | 849 | 80 | |
AK135194.1 | - | 11 - 12 | 1457 | 80 | |
AK135353.1 | - | 11 - 12 | 1457 | 80 | |
AK145707.1 | - | 11 - 12 | 1482 | 80 | |
AK159717.1 | - | 11 - 12 | 1459 | 80 | |
AK159824.1 | - | 11 - 12 | 1457 | 80 | |
AK159937.1 | - | 12 - 13 | 1433 | 80 | |
AK166191.1 | - | 11 - 12 | 1457 | 80 | |
AK168410.1 | - | 11 - 12 | 1460 | 80 | |
AK168840.1 | - | 11 - 12 | 1457 | 80 | |
AK170610.1 | - | 11 - 12 | 1457 | 80 | |
AK172336.1 | - | 11 - 12 | 1464 | 80 | |
BC048181.1 | - | 11 - 12 | 1444 | 80 | |
BC098502.1 | - | 11 - 12 | 1433 | 80 | |
X60671.1 | - | 11 - 12 | 1439 | 80 |