The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048167.1 | NP_001041632.1 | 2 - 3 | 2082 | 87 |
NM_010837.3 | NP_034967.2 | 2 - 3 | 2082 | 87 | |
GenBank mRNA | AK018193.2 | - | 2 - 3 | 1562 | 87 |
AK087413.1 | - | 2 - 3 | 2045 | 87 | |
BC053039.1 | - | 2 - 3 | 2083 | 87 |