The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011078.3 | NP_035208.2 | 17 - 18 | 2457 | 69 |
GenBank mRNA | AB221590.1 | - | 6 - 7 | 843 | 69 |
AF043726.1 | - | 17 - 18 | 2457 | 69 | |
AK134358.1 | - | 3 - 4 | 285 | 69 | |
AK172995.1 | - | 15 - 16 | 2164 | 69 | |
BC051633.1 | - | 17 - 18 | 2455 | 69 |