The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040691.1 | NP_001035781.1 | 4 - 5 | 582 | 93 |
NM_011677.2 | NP_035807.2 | 3 - 4 | 554 | 93 | |
GenBank mRNA | AK146178.1 | - | 4 - 5 | 582 | 93 |
AK168709.1 | - | 3 - 4 | 504 | 93 | |
AK168785.1 | - | 3 - 4 | 504 | 93 | |
BC004037.1 | - | 2 - 3 | 384 | 93 | |
BC011039.1 | - | 4 - 5 | 639 | 93 | |
BC052853.1 | - | 3 - 4 | 498 | 93 | |
U55041.1 | - | 3 - 4 | 508 | 93 | |
X99018.1 | - | 3 - 4 | 510 | 93 | |
Y08975.1 | - | 4 - 5 | 582 | 93 |