The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013750.2 | NP_038778.1 | 1 - 2 | 810 | 128 |
GenBank mRNA | AK003767.1 | - | 1 - 2 | 776 | 128 |
AK199905.1 | - | 1 - 2 | 110 | 128 | |
AK206046.1 | - | 1 - 2 | 106 | 128 | |
AK210974.1 | - | 1 - 2 | 104 | 128 | |
AK214255.1 | - | 1 - 2 | 104 | 128 | |
BC023376.1 | - | 1 - 2 | 712 | 128 | |
BC023408.1 | - | 1 - 2 | 712 | 128 |