The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013791.2 | NP_038819.1 | 6 - 7 | 734 | 79 |
GenBank mRNA | AK037967.1 | - | 6 - 7 | 735 | 79 |
AK077541.1 | - | 6 - 7 | 730 | 79 | |
AK136123.1 | - | 3 - 4 | 417 | 79 | |
BC013703.1 | - | 6 - 7 | 718 | 79 | |
U72194.1 | - | 6 - 7 | 717 | 79 |