Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mkln1
Entrez Gene ID
27418
Gene Name
muskelin 1, intracellular mediator containing kelch motifs
Gene Aliases
A130067F06Rik, AU015903, BB154892
Chromosome Location
Chr.6: 31398828 - 31511795 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013791.2 NP_038819.1 13 - 14 1704 148
GenBank mRNA BC013703.1 - 13 - 14 1688 148
U72194.1 - 13 - 14 1687 148

View Full Product Details