The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252580.1 | NP_001239509.1 | 18 - 19 | 3317 | 103 |
NM_019729.3 | NP_062703.2 | 18 - 19 | 3284 | 103 | |
GenBank mRNA | AB045709.1 | - | 17 - 18 | 3058 | 103 |
AF057146.1 | - | 17 - 18 | 3075 | 103 | |
AK122195.1 | - | 18 - 19 | 3195 | 103 | |
AK129003.1 | - | 18 - 19 | 3285 | 103 | |
AK134058.1 | - | 18 - 19 | 3239 | 103 | |
AK142881.1 | - | 15 - 16 | 2782 | 103 | |
AK153015.1 | - | 17 - 18 | 3031 | 103 | |
AK166540.1 | - | 16 - 17 | 2638 | 103 | |
BC027052.1 | - | 8 - 9 | 1837 | 103 | |
BC050947.1 | - | 18 - 19 | 3249 | 103 | |
BC061465.1 | - | 18 - 19 | 3249 | 103 | |
BC066126.1 | - | 18 - 19 | 3247 | 103 | |
BC092078.1 | - | 6 - 7 | 1077 | 103 |