The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190846.1 | NP_001177775.1 | 7 - 8 | 901 | 63 |
NM_020005.4 | NP_064389.2 | 7 - 8 | 1455 | 63 | |
GenBank mRNA | AF254442.1 | - | 7 - 8 | 1096 | 63 |
AK030070.1 | - | 7 - 8 | 902 | 63 | |
AK132486.1 | - | 7 - 8 | 1155 | 63 | |
AK156290.1 | - | 7 - 8 | 1455 | 63 | |
BC082581.1 | - | 7 - 8 | 1111 | 63 | |
BC138195.1 | - | 7 - 8 | 1123 | 63 | |
BC145896.1 | - | 7 - 8 | 1123 | 63 |