The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168346.1 | NP_001161818.1 | 6 - 7 | 2120 | 73 |
NM_023699.3 | NP_076188.3 | 6 - 7 | 2120 | 73 | |
GenBank mRNA | AK159078.1 | - | 6 - 7 | 1966 | 73 |
EU887656.1 | - | 6 - 7 | 1878 | 73 | |
EU887657.1 | - | 6 - 7 | 1878 | 73 | |
EU887658.1 | - | 1 - 2 | 333 | 73 | |
EU887659.1 | - | 1 - 2 | 333 | 73 |