The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163645.1 | NP_001157117.1 | 3 - 4 | 678 | 70 |
NM_027881.3 | NP_082157.2 | 2 - 3 | 288 | 70 | |
GenBank mRNA | AK004768.1 | - | 2 - 3 | 290 | 70 |
AK037114.1 | - | 2 - 3 | 288 | 70 | |
AK160310.1 | - | 3 - 4 | 679 | 70 | |
AK173006.1 | - | 3 - 4 | 487 | 70 | |
BC044866.1 | - | 2 - 3 | 323 | 70 | |
BC086485.1 | - | 3 - 4 | 590 | 70 |