Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcnxl3
Entrez Gene ID
104401
Gene Name
pecanex-like 3 (Drosophila)
Gene Aliases
mKIAA4073
Chromosome Location
Chr.19: 5664635 - 5688908 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144868.3 NP_659117.2 25 - 26 4760 79
GenBank mRNA AF237953.1 - 23 - 24 2777 79
AK220471.1 - 3 - 4 658 79
BC038056.1 - 20 - 21 2868 79
BC054751.1 - 25 - 26 4039 79
BC055289.1 - 26 - 27 4563 79
BC096538.1 - 25 - 26 4760 79

View Full Product Details