The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024193.2 | NP_077155.2 | 6 - 7 | 821 | 100 |
GenBank mRNA | AK009799.1 | - | 6 - 7 | 807 | 100 |
AK011481.1 | - | 6 - 7 | 788 | 100 | |
AK077795.1 | - | 6 - 7 | 788 | 100 | |
AK139924.1 | - | 6 - 7 | 788 | 100 | |
AK146243.1 | - | 6 - 7 | 788 | 100 | |
AK150258.1 | - | 6 - 7 | 788 | 100 | |
BC002231.1 | - | 1 - 2 | 173 | 100 | |
BC021355.1 | - | 6 - 7 | 783 | 100 |